Kinesis® SureStop® Short Thread Glass Vials, 9 mm
Optimum vial geometry provides trouble-free autosampler operation
  • No more over tightening and deforming of septa
  • Extend autosampler needle lifetime
SureStop vials feature specially designed over-wind barrier to prevent the cap from being overtightened, ensuring that the septum will not be deformed, minimizing damage to the autosampler needles and preventing sample evaporation. SureStop vials are compatible with all short thread caps and septa.
28,200.00 - ₹32,900.00INR / Pkg of 1000
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Special Features Tube Color Volume (mL)
Availability Pricing
COMPARE
Label and filling lines Amber 2
₹32,900.00INR / Pkg of 1000
COMPARE
Label and filling lines Clear 2
₹30,500.00INR / Pkg of 1000
COMPARE
N/A Clear 2
₹28,200.00INR / Pkg of 1000