Laser Pocket Tachometers
Ergonomic design allows simultaneous viewing of both the target and display
  • Rugged, rubberized design for industrial applications
These digital, portable tachometers provide a nonslip, rubber grip for single-hand operation. Tachometers feature user-selectable fixed or auto-ranging resolution, memory of last reading, minimum and maximum, tripod mounting bushing for hands-free operation, target “lock on” indicator, and a low-battery indicator.
Model 08199-20 features a red LED that measures optical rpm from up to 30" (76.2 cm) away.
Models 08199-22 and –24 feature a Class II laser that measures rpm from up to 25 feet (7.6 m) away. These tachometers function as a totalizer/counter and a timer (stopwatch) programmable in both English and metric units. Model 08199-24 adds the functionality of measuring contact rpm and surface speed.
45,400.00 - ₹95,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Display Type Max RPM Range (rpm) Description
Availability Pricing
Monarch 6109-010 Optical/LED Pocket Tachometer 0819920 Mfr # 6109-010
COMPARE
5-digit alphanumeric LCD, 0.5" H (1.3 cm) 99999
₹45,400.00INR / Each
Monarch PLT-200 Optical/Laser Pocket Tachometer 0819922 Mfr # 6125-010
COMPARE
5-digit alphanumeric LCD, 0.5" H (1.3 cm) 200000
₹60,700.00INR / Each
Monarch PLT200KIT Laser/Contact Pocket Tachometer Kit with RCA Kit 0819924 Mfr # 6125-011
COMPARE
5-digit alphanumeric LCD, 0.5" H (1.3 cm) 200000
₹95,000.00INR / Each