Oakton® SALT 6+ Handheld Salinity Meters
Autoranging salinity with temperature measurements
  • Temperature readout in °C with the push of a button
  • Rubber boot with built-in stand protects meter and adds functionality
These microprocessor-based meters feature easy, push-button operation that offers high accuracy at an extremely affordable price. The SALT 6+ features single-point calibration; calibrate using 4487-ppm calibration solution 00653-89 (sold separately).
Additional features include a convenient Hold function and auto-off function that shuts meter off after 20 minutes of nonuse. Programming functions allow customization of meter for temperature compensation, temperature coefficient, and normalization for better accuracy in varying applications.
Choose from meter with probe or meter with probe kit. The kit adds calibration solutions, sample bottles, rinse bottle, and hard plastic carrying case.
110,000.00 - ₹157,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Handheld Salinity Meter with Probe
₹157,000.00INR / Each
COMPARE
Handheld Salinity Meter with Probe and NIST-Traceable Calibration
₹117,000.00INR / Each
COMPARE
Handheld Salinity Meter Kit
₹110,000.00INR / Each
COMPARE
Handheld Salinity Meter Kit with NIST-Traceable Calibration
₹131,000.00INR / Each