Oakton® TDS 6+ Handheld TDS Meters
Autoranging conductivity or TDS with temperature measurement
  • TDS 6+ meters have five auto-ranging measurement modes—up to 200.0 mS (200.0 ppt)
  • Temperature readout in °C with the push of a button
  • Rubber boot with built-in stand protects meter and adds functionality
These microprocessor-based meters feature easy, push-button operation that offers high accuracy at an extremely affordable price. A quick and easy single-point calibration covers all five ranges with the TDS 6+ meters; for greater precision, single points can be calibrated within each of the five measurement ranges.
Additional features include a convenient Hold function and auto-off function that shuts meter off after 20 minutes of nonuse. Programming functions allow customization of meter for temperature compensation, temperature coefficient, normalization, cell constant, and conductivity-to-TDS conversion factor for better accuracy in varying applications.
96,400.00 - ₹131,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
handheld TDS meter with probe
₹96,400.00INR / Each
COMPARE
handheld TDS meter with probe and NIST-traceable Calibration
₹117,000.00INR / Each
COMPARE
Handheld Meter Kit
₹110,000.00INR / Each
COMPARE
handheld TDS meter kit with NIST-traceable Calibration
₹131,000.00INR / Each