PYREX Brand 4320 Boiling Flasks
Boiling, round bottom, short neck, joint
These boiling flasks with short neck are carefully designed and manufactured from borosilicate glass. Uniform wall thickness, a characteristic of all round-bottom, Pyrex glassware, provides a balance between expansion and mechanical strength for greater shock resistance and versatility. Use boiling flasks for a variety of mixing, heating, and boiling applications commonly encountered in most laboratory work. The normal working temperature is 230°>C. Includes a Ts joint.
12,100.00 - ₹230,000.00INR / Pkg of 1
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Joint Size Number Material
Availability Pricing
Pyrex 4320-500 Brand 4320 round-bottom flask; 500 mL, pack of 2 3455304 Mfr # 4320-500
COMPARE
500 24/40 Glass
₹20,400.00INR / Pkg of 2
Pyrex 4320-1L Brand 4320 round-bottom flask; 1000 mL, pack of 1 3455305 Mfr # 4320-1L
COMPARE
1000 24/40 Glass
₹12,100.00INR / Pkg of 1
Pyrex 4320-2L Brand 4320 round-bottom flask; 2000 mL, pack of 1 3455306 Mfr # 4320-2L
COMPARE
2000 24/40 Glass
₹15,800.00INR / Pkg of 1
Pyrex 4320-100 Brand 4320 round-bottom flask; 100 mL, case of 12 3455322 Mfr # 4320-100
COMPARE
100 24/40 Glass
₹105,000.00INR / Case of 12
Pyrex 4320-250 Brand 4320 round-bottom flask; 250 mL, case of 12 3455323 Mfr # 4320-250
COMPARE
250 24/40 Glass
₹114,000.00INR / Case of 12
Pyrex 4320-500 Brand 4320 round-bottom flask; 500 mL, case of 12 3455324 Mfr # 4320-500
COMPARE
500 24/40 Glass
₹122,000.00INR / Case of 12
Pyrex 4320-1L Brand 4320 round-bottom flask; 1000 mL, case of 12 3455325 Mfr # 4320-1L
COMPARE
1000 24/40 Glass
₹145,000.00INR / Case of 12
Pyrex 4320-2L Brand 4320 round-bottom flask; 2000 mL, case of 6 3455326 Mfr # 4320-2L
COMPARE
2000 24/40 Glass
₹119,000.00INR / Case of 6
Pyrex 4320-5L Brand 4320 round-bottom flask; 5000 mL, case of 4 3455327 Mfr # 4320-5L
COMPARE
5000 45/50 Glass
₹230,000.00INR / Case of 4
Pyrex 4320-12L Brand 4320 round-bottom flask; 12000 mL, case of 1 3455328 Mfr # 4320-12L
COMPARE
12000 55/50 Glass
₹108,000.00INR / Case of 1
Showing 1- 10 of 10