Rubbermaid Storage Containers with Handles, HDPE
Label these containers on the handy "date-content" area on the lid to keep track of contents
  • Graduated in quarts and liters
  • NSF certified
White high-density polyethylene (HDPE) storage containers with handles are dishwasher safe and withstand temperatures from -40 to 150°>F (-40 to 66°>C). Shoulders facilitate stacking and the rounded corners clean up quickly. Lids are sold separately.
13,000.00 - ₹57,500.00INR / Pkg of 6
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Material Description
Availability Pricing
Rubbermaid 5720 WHITE. Storage Containers with Handle, White HDPE, 2 quart 6351101 Mfr # 5720 WHITE.
COMPARE
2 HDPE Storage Containers with Handle, White HDPE, 2 quart
₹13,000.00INR / Pkg of 12
Rubbermaid 5721 WHITE Storage Containers with Handle, White HDPE, 4 quart 6351103 Mfr # 5721 WHITE
COMPARE
4 HDPE Storage Containers with Handle, White HDPE, 4 quart
₹17,800.00INR / Pkg of 12
Rubbermaid 5723 WHITE Storage Containers with Handle, White HDPE, 6 quart 6351105 Mfr # 5723 WHITE
COMPARE
6 HDPE Storage Containers with Handle, White HDPE, 6 quart
₹29,400.00INR / Pkg of 12
Rubbermaid 5724 WHITE Storage Containers with Handle, White HDPE, 8 quart 6351107 Mfr # 5724 WHITE
COMPARE
8 HDPE Storage Containers with Handle, White HDPE, 8 quart
₹23,300.00INR / Pkg of 12
Rubbermaid 5726 WHITE Storage Containers with Handle, White HDPE, 12 quart 6351109 Mfr # 5726 WHITE
COMPARE
12 HDPE Storage Containers with Handle, White HDPE, 12 quart
₹32,600.00INR / Pkg of 6
Rubbermaid 5729 WHITE Storage Containers with Handle, White HDPE, 22 quart 6351113 Mfr # 5729 WHITE
COMPARE
22 HDPE Storage Containers with Handle, White HDPE, 22 quart
₹57,500.00INR / Pkg of 6