Specific Gravity Cups
Quickly determine specific gravity or weight per gallon
  • Cups with tare weights let you use your pan balance for specific gravity measurements
  • Tare weights are adjusted to within ±0.2 grams of cup weight (0.1 gram for 8.32-mL cups)
Use these precision-made stainless steel cups with your laboratory balance to determine specific gravity or pounds per gallon. Simply tare balance to cup weight, then weigh cup filled with sample. Order cups for U.S. Standard or British Standard determinations. All cups meet MIL STD 45662A and are NIST-traceable.
71,700.00 - ₹228,000.00INR / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Height (cm) Weight (With Water) (g)
Availability Pricing
BYK-Gardner PV-9654 Specific Gravity Cup, 83.2 mL, 0.5% Tolerance, 76 mm H x 38 mm OD; U.S. use 0871190 Mfr # PV-9654
COMPARE
83.2 3.8 213
₹71,700.00INR / Each
BYK-Gardner PV-9664 Specific Gravity Cup, 8.32 mL, 1.0% Tolerance; U.S. use 0871192 Mfr # PV-9664
COMPARE
8.32 2.5 47
₹90,600.00INR / Each
Gardco WG-SS-8.32 Specific Gravity Cup, 8.32 mL, 1.2% Tolerance; U.S. use 3800000 Mfr # WG-SS-8.32
COMPARE
8.32 2.65 47
₹78,000.00INR / Each
Gardco WG-SS-8.32/T Specific Gravity Cup w/Tare Weight, 8.32 mL, 1.2% Tolerance; U.S. use 3800002 Mfr # WG-SS-8.32/T
COMPARE
8.32 2.65 47
₹101,000.00INR / Each
Gardco WG-SS-8.32/C Specific Gravity Cup with NIST Calibration Certificate, 8.32 mL, 1.2% Tolerance; for US-Use 3800004 Mfr # WG-SS-8.32/C
COMPARE
8.32 2.65 47
₹149,000.00INR / Each
Gardco WG-SS-8.32/T/C Specific Gravity Cup with Tare Weight and NIST Calibration Certificate, 8.32 mL, 1.2% Tol; US-Use 3800006 Mfr # WG-SS-8.32/T/C
COMPARE
8.32 2.65 47
₹200,000.00INR / Each
Gardco WG-SS-83.2 Specific Gravity Cup, 83.2 mL, 0.5% Tolerance, 76.2 x 38.1 mm OD; U.S. use 3800010 Mfr # WG-SS-83.2
COMPARE
83.2 7.62 213
₹80,200.00INR / Each
Gardco WG-SS-83.2/T Specific Gravity Cup w/Tare Weight, 83.2 mL, 0.5% Tolerance; U.S. use 3800012 Mfr # PV-9655
COMPARE
83.2 7.6 213
₹108,000.00INR / Each
Gardco WG-SS-83.2/C Specific Gravity Cup with NIST Calibration Certificate, 83.2 mL, 0.5% Tolerance; US-Use 3800014 Mfr # WG-SS-83.2/C
COMPARE
83.2 6.35 213
₹152,000.00INR / Each
Gardco WG-SS-83.2/T/C Specific Gravity Cup with Tare Weight and NIST Calibration Certificate, 83.2 mL, 0.5% Tol; US-Use 3800016 Mfr # WG-SS-83.2/T/C
COMPARE
83.2 6.35 213
₹206,000.00INR / Each
Gardco WG-SS-100 Specific Gravity Cup, 100 mL Capacity, 0.5% Volume Tolerance, for British use 3800020 Mfr # WG-SS-100
COMPARE
100 7.83 243
₹106,000.00INR / Each
Gardco WG-SS-100/T Specific Gravity Cup w/Tare Weight, 100 mL; British use 3800022 Mfr # WG-SS-100/T
COMPARE
100 7.83 243
₹132,000.00INR / Each
Gardco WG-SS-100/C Specific Gravity Cup with NIST Calibration Certificate, 100 mL; British-Use 3800024 Mfr # WG-SS-100/C
COMPARE
100 7.83 243
₹177,000.00INR / Each
Gardco WG-SS-100/T/C Specific Gravity Cup with Tare Weight and NIST Calibration Certificate, 100 mL; British-Use 3800026 Mfr # WG-SS-100/T/C
COMPARE
100 7.83 243
₹228,000.00INR / Each
Showing 1- 14 of 14