Thermo Scientific™ SuperNuova+™ Digital Hot Plates
Don't wait for long warm-up times
  • Heat to the maximum temperature in 5 minutes for 7" x 7" models, and 7 minutes for 10" x 10" models
  • Program and save temperature and time settings for your most common applications
  • New design features a large two-line LED display raised to protect from splashes and spills
  • Temperature stability of ±1.8°F (1°C) gives a reliable, repeatable performance
  • Red "hot top" light provides warning once the top plate temperature rises above 122°F (50°C) to prevent accidental burns
Control the temperature using the internal type K thermocouple sensor for standard applications, or externally using the stainless steel 100 Pt RTD probe (included) to maintain a more precise set point. The two-line LED display shows both set and actual temperatures; if using the remote probe, "R" will display after the actual temperature. Program and save up to three heat and time settings. Use the presets individually for automatic shut-off, or use the Preset Continue function to run the programs continuously for ramp/dwell control.
The newly designed display is tilted to be easily read from above, and separated and raised to protect it from splashes and spills. A red, flashing LED light warns you when the top plate temperature is above 122°F (50°C) for added safety.
292,000.00 - ₹386,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Top Plate Length (in) Top Plate Material
Availability Pricing
Thermo Scientific SuperNuova+ HP88857195 SuperNuova+ Hot Plate, 7x7" Ceramic; 230 VAC 0460026 Mfr # HP88857195
₹292,000.00INR / Each
COMPARE
130 7 1/4 Ceramic
Loading
₹292,000.00INR / Each
Thermo Scientific SuperNuova+ HP88857198 Super-Nuova+ Hot Plate, 7x7" Aluminum; 230 VAC 0460033 Mfr # HP88857198
₹369,000.00INR / Each
COMPARE
230 7 1/4 Aluminum
Loading
₹369,000.00INR / Each
Thermo Scientific SuperNuova+ HP88850195 Super-Nuova+ Hot Plate, 10x10" Ceramic; 230 VAC 0460051 Mfr # HP88850195
₹386,000.00INR / Each
COMPARE
230 10 1/2 Ceramic
Loading
₹386,000.00INR / Each