Troemner Class-4 Electronic Balance Calibration Weights
ASTM Class 4 - For calibration of entry level balances and for student use.
  • Two-piece weights with adjusting cavities
  • Manufactured from stainless-steel chosen for its uniformity density, high resistance to wear and corrosion, and low magnetic properti
Electronic Balance Weights can be used to check or verify the accuracy of your balance and/or scale between calibrations. It is recommended that the user check a balance and/or scale prior to each use. Keeping the weight at or near the balance and/or scale keeps the weight at the same temperature as the equipment.To get all the accuracy your electronic balance and/or scale provides, you need to perform routine calibrations and verifications using precision calibration weights. Troemner's Electronic Balance Weights are intended for this purpose. Electronic Balance Weights are ideal for non chemical environments to verify balances or scales.Manufactured from stainless-steel chosen for its uniformity density, high resistance to wear and corrosion, and low magnetic properties. These individual stainless steel electronic balance masses are designed for calibration of electronic balances and routine weighing applications. The larger denomination electronic balance calibration weights (5 kg and up) have a recessed stainless steel grip handle for easy handling and stacking. Smaller denominations (2000 g and lower) are cylindrical in shape with the heavier of these having a groove near the top for easy gripping.
26,800.00 - ₹447,000.00INR / Each
33 variations of this product are available.
See All Product Options Below
33 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mass Certification Mass Size (g) Description
Availability Pricing
Troemner 8458W 10g Electronic Class 4 Weight with NVLAP Accredited Certificate 0102294 Mfr # 8458W
COMPARE
NVLAP 10
₹34,400.00INR / Each
Troemner 8452W 50g Electronic Class 4 Weight with NVLAP Accredited Certificate 0102296 Mfr # 8452W
COMPARE
NVLAP 50
₹34,400.00INR / Each
Troemner 8416W 5kg Electronic Class 4 Weight with NVLAP Accredited Certificate 0102297 Mfr # 8416W
COMPARE
NVLAP 5000
₹151,000.00INR / Each
Troemner 8400T 30 kg Class 4 Electronic Balance Weight with Traceable Cert 0103370 Mfr # 8400T
COMPARE
NVLAP 30000
₹447,000.00INR / Each
Troemner 8456T 20 g Class 4 Electronic Balance Weight with Traceable Cert 0103373 Mfr # 8456T
COMPARE
NIST 20
₹26,800.00INR / Each
Troemner 8436W 300g, Electronic Class 4 Weight with NVLAP Certificate 0910803 Mfr # 8436W
COMPARE
NVLAP 300
₹49,800.00INR / Each
Troemner 8410W 10kg, Electronic Class 4 Weight with NVLAP Certificate 0910873 Mfr # 8410W
COMPARE
NVLAP 10000
₹224,000.00INR / Each
Troemner 8414W 5kg, Electronic Class 4 Weight with NVLAP Certificate 0910875 Mfr # 8414W
COMPARE
NVLAP 5000
₹162,000.00INR / Each
Troemner 8420W 4kg, Electronic Class 4 Weight with NVLAP Certificate 0910876 Mfr # 8420W
COMPARE
NVLAP 4000
₹144,000.00INR / Each
Troemner 8422W 3kg, Electronic Class 4 Weight with NVLAP Certificate 0910877 Mfr # 8422W
COMPARE
NVLAP 3000
₹98,500.00INR / Each
Troemner 8424W 2kg, Electronic Class 4 Weight with NVLAP Certificate 0910878 Mfr # 8424W
COMPARE
NVLAP 2000
₹70,700.00INR / Each
Troemner 8428W 1000g, Electronic Class 4 Weight with NVLAP Certificate 0910880 Mfr # 8428W
COMPARE
NVLAP 1000
₹61,900.00INR / Each
Troemner 8432W 500g, Electronic Class 4 Weight with NVLAP Certificate 0910881 Mfr # 8432W
COMPARE
NVLAP 500
₹55,900.00INR / Each
Troemner 8434W 400g, Electronic Class 4 Weight with NVLAP Certificate 0910882 Mfr # 8434W
COMPARE
NVLAP 400
₹52,000.00INR / Each
Troemner 8438W 200g, Electronic Class 4 Weight with NVLAP Certificate 0910883 Mfr # 8438W
COMPARE
NVLAP 200
₹49,100.00INR / Each
Troemner 8442W 150g, Electronic Class 4 Weight with NVLAP Certificate 0910884 Mfr # 8442W
COMPARE
NVLAP 150
₹53,700.00INR / Each
Troemner 8444W 100g, Electronic Class 4 Weight with NVLAP Certificate 0910885 Mfr # 8444W
COMPARE
NVLAP 100
₹36,200.00INR / Each
Troemner 8458T 10g, Electronic Class 4 Weight with NVLAP Certificate 1195605 Mfr # 8458T
COMPARE
NVLAP 10
₹26,800.00INR / Each
Troemner 8454T 30g, Electronic Class 4 Weight with NVLAP Certificate 1195610 Mfr # 8454T
COMPARE
NVLAP 30
₹26,800.00INR / Each
Troemner 8452T 50g, Electronic Class 4 Weight with NVLAP Certificate 1195615 Mfr # 8452T
COMPARE
NVLAP 50
₹28,700.00INR / Each
Showing 1 - 20 of 33