DWK Life Sciences (Kimble) Plastic-Coated HPLC Reservoirs without Graduation, 5000 mL
Representative image only.
DWK Life Sciences (Kimble) Mfr # 953901-5002 – Item # 1595875
Specifically designed for the preparation, storage and delivery of all liquid chromatography mobile phases.
  • Manufactured from the most chemically inert glass to prevent the leaching of any extractables into the mobile phase solvents
  • PVC plastic coating blocks virtually all UV light up to 385 nm, preventing photo-degradation of light-sensitive reagents
  • Recommended for use under vacuum for filtration and vacuum degassing of mobile phase solvents
  • Can also be used at internal pressures up to 6 psig for helium sparging and blanketing of the mobile phase
  • Conical bottom reservoirs deliver virtually all of the mobile phase without dangerous reservoir tilting
  • Patent No. Des. 292,824
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
PRODUCT OPTIONS
₹61,800.00INR / Case
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPlastic-Coated HPLC Reservoirs without Graduation, 5000 mL
Key Features
  • Manufactured from the most chemically inert glass to prevent the leaching of any extractables into the mobile phase solvents
  • PVC plastic coating blocks virtually all UV light up to 385 nm, preventing photo-degradation of light-sensitive reagents
  • Recommended for use under vacuum for filtration and vacuum degassing of mobile phase solvents
  • Can also be used at internal pressures up to 6 psig for helium sparging and blanketing of the mobile phase
  • Conical bottom reservoirs deliver virtually all of the mobile phase without dangerous reservoir tilting
  • Patent No. Des. 292,824
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements