Temperature Compatibility Guidelines for Centrifuge Tubes and Bottles
Temperature Compatibility Guidelines
for Centrifuge Tubes and Bottles
Material Nonrefrigerated Refrigerated Autoclavable*
Polypropylene (PP) Strong and rigid;
holds shape and performs well,
even at high speeds
Not recommended; becomes
brittle and may craze or
crack at high speeds
Yes
Polypropylene
copolymer (PPCO)
Less rigid; closures are
required at high speeds
Rigid but not brittle;
optimal for high speeds
Yes
Polycarbonate (PC)
and polysulfone (PSF)
Retains rigidity and mechanical
strength, even at high speeds
Retains rigidity and mechanical
strength, even at high speeds
Yes
PTFE Flexible; can be used at low
speeds; refrigeration preferred
at high speeds
Excellent for high speeds;
tubes must be run full and
tightly capped
Yes
Tefzel® ETFE Lacks rigidity;
closures required for
added support at high speeds
Excellent for high speeds; closures
required for added support
Yes
Low-density
polyethylene (LDPE)
Flexible; use at low to
medium speeds
More rigid;
excellent for high speeds
No
High-density
polyethylene (HDPE)
Use at low speeds only Use at low speeds only No
Polymethylpentene
(PMP)
Somewhat brittle; use at
low speeds only
Somewhat brittle; use at
low speeds only
Yes
Polystyrene (PS) Use at low speeds only Use at low speeds only No
*Note: Before autoclaving, set closure on top of tube without engaging threads.