pH Temperature Compensation
pH Temperature Compensation
Typical pH electrode response
as a function of temperature
The pH of any solution is a function of its temperature. Voltage output from the electrode changes linearly in relationship to changes in pH, and the temperature of the solution determines the slope of the graph. One pH unit corresponds to 59.16 mV at 25°C, the standard voltage and temperature to which all calibrations are referenced. The electrode voltage decreases to 54.20 mV/pH unit at 0.0°C and increases to 74.04 mV/pH unit at 100.0°C (see graph).
Since pH values are temperature dependent, pH applications require some form of temperature compensation to ensure standardized pH values. Meters and controllers with automatic temperature compensation (ATC) receive a continuous signal from a temperature element and automatically correct the pH value based on the temperature of the solution. Manual temperature compensation requires the user to enter the temperature. ATC is considered to be more practical for most pH applications.