Fitting MaterialPolysulfone with Polycarbonate Sleeve
Sterilization MethodFull connector assembly: gamma irradiation up to 50 kGy or autoclave up to 129°C (265°F) for 60 minutes Steam-in-Place (SIP): up to 130°C (266°F) for 60 minutes
DescriptionSanitary Steam-in-Place Connector, Two Cycle, Polysulfone with Polycarbonate Sleeve, 1/2" ID x 3/4" x 3/4" Sanitary; 1/Ea
Key Features
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
Free technical application support available to determine which connector type is needed for your application
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Innovative three-port steam design
Offers both “steam on” and “steam off” functionalities
Steam-Thru II thumb latch secures valve position
3/4" sanitary connection for condensate port
0.5" nominal flow path size
1/2" hose barb x 3/4" sanitary connection
More About this Item
The CPC® (Colder) Steam-Thru® II connectors feature a unique three-port design for a true steam-through steam-in-place (SIP) process, eliminating dead legs and the need for a laminar flow hood. Use the Steam-Thru II SIP connection to sterilize the fitting while connected to stainless steel equipment. The Steam-Thru II connection offers the flexibility of “steam on” and “steam off” functionality, allowing a second SIP cycle following media transfer. The “steam off” function helps minimize cross-contamination risks associated with reusable components. Select from 3/4" and 1-1/2" sanitary connections for attachment to the processing equipment. Connectors withstand temperatures up to 135°C (275°F) for 60 minutes during SIP.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, cell therapy, gene therapy, and other critical-use applications. Fittings are manufactured, double bagged, and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.