High-Accuracy Rugged Tachometers
Rubber boot protects meter during industrial applications
  • Provides ±0.02% accuracy with 0.01-rpm resolution
These digital tachometers feature high accuracy for precision measurements. The removable rubber boot provides an ergonomic, comfortable grip while protecting the meter in heavy-duty environments. Tachometers are available in optical (noncontact) or optical/contact models. Memory stores min/max, average, and last readings during measurement. A red LED helps target equipment from up to 24" (61.0 cm) away. Model 87601-03 features optical/contact measurements in rpm, three surface speed ranges, and three length measurement ranges.
54,300.00 - ₹80,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Display Type Max RPM Range (rpm)
Availability Pricing
Electro Numerics CDT-1000HD-N High-Accuracy Rugged Noncontact Optical Tachometer 8760101 Mfr # CDT-1000HD-N
COMPARE
5-digit LCD, 0.375" H (1.0 cm) 99999
₹54,300.00INR / Each
Electro Numerics CDT-2000HD-N High-Accuracy Rugged Contact/Noncontact Optical Tachometer 8760103 Mfr # CDT-2000HD-N
COMPARE
5-digit LCD, 0.375" H (1.0 cm) 99999
₹80,600.00INR / Each