Megger TTRU1 Handheld Transformer Turns Ratiometer (TTR) Test Sets
Specially designed to test distribution, instrument (CTs and PT/VTs), and power transformers
  • Perform routine polarity validation and turns ratio tests
  • Easy-to-use interface on full color bright LCD
  • Click start and perform over 1000 turns ratio tests on a battery charge!
  • Pass/fail ratio evaluation
  • Additive/Subtractive polarity validation
  • Excitation current measurements
  • Export saved results for analysis
The TTRU1 handheld meters use a single-phase step-down method to measure the ratio and polarity of the transformer's turns. It can help identify any shorted turns, open-circuited conditions, or insulation deterioration to ensure the power transformer is performing properly.
A full-color, hi-bright LCD screen, rotary knob, seven function buttons, and an easy-to-follow setup provide the right result every time. The meter performs a vector group validation during every test to determine whether the transformer is additive or subtractive. Use the excitation current test to locate defects in magnetic core balance, magnetic core structure, shifting windings, failures in the turn-to-turn insulation, or problems in tap changers.
Store up to 10,000 sets of single-phase results on the meter. Install PC software to view and download data to a computer or a thumb drive via the USB-A connection. The meter operates on six NiMH rechargeable batteries, offering maximum portability and flexibility; it performs up to 1000 TTR tests on a single charge. Simply charge batteries by connecting to a wall charger or computer USB port.
1,141,000.00 - ₹1,310,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
AC Max (Volts) Ac Voltage Accuracy Description
Availability Pricing
Megger TTRU1 Basic Transformer Turns Ratiometer Test Set, 37.5 VAC Max Induced 2002730 Mfr # TTRU1-BASIC
COMPARE
37.5 ±0.05% from -20 to 50 °C (-4 to 122 °F)
₹1,141,000.00INR / Each
Megger TTRU1 Advanced Transformer Turns Ratiometer Test Set, 62.5 VAC Max Induced 2002731 Mfr # TTRU1-ADV
COMPARE
62.5 ±0.05% from -20 to 50 °C (-4 to 122 °F)
₹1,310,000.00INR / Each